Learn More
DWK Life Sciences Kimble™ KIMAX™ Soxhlet Apparatus with Allihn Condenser, Condenser only
Catalog No. p-145637
Change view
Click to view available options
Standard Taper Joint:
34/45
45/50
55/50
Diameter (Metric) Inside:
3 cm
4 cm
5 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint Diameter (Metric) Inside
0955110B 45/50 4 cm
0955110A 34/45 3 cm
0955110C 55/50 5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0955110B Supplier DWK Life Sciences Supplier No. 2402740
Only null left
For use with for Kimble™ extraction apparatus
  • The soxhlet extractor was originally designed for the extraction of a lipid from a solid material
  • Not limited to the extraction of lipids - typically, a soxhlet extraction is only required where the desired compound has a limited solubility in a solvent and the impurity is insoluble in that solvent
  • If the desired compound has a high solubility in a solvent then a simple filtration can be used to separate the compound from the insoluble substance
Order Info
See the Complete Assembly (09-551A, 09-551B, 09-551C) which contains a condenser, extraction tube and flask.
Specifications
Product Type Extractor Condenser
For Use With (Equipment) Soxhlet Extraction Apparatus
Standard Taper Joint 45/50
Diameter (English) Inside 1.57 in.
Diameter (Metric) Inside 4 cm