missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ Plastic Safety-Coated Class A Volumetric Flasks with Glass Stopper
Heavy-duty, wide-mouth flasks
Supplier: DWK Life Sciences KC92812G4000
Description
Heavy-duty, wide-mouth flasks
KIMAX KimCote protective glassware coating goes beyond traditional coatings. Should a break occur, KimCote will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. It's ultra-clear, extremely durable, autoclavable and resistant to many common laboratory chemicals. KimCote's unique texture also provides a non-slip handling surface, wet or dry. Flasks are graduated to Class A volumetric tolerances and are designed from ASTM Specification E288. Glass stoppers are included.- Neck dimensions will easily accommodate a 50 mL pipet
- Superior strength, durability and heavy uniform walls ensure safety in the lab
- Flat interior bottoms are ideal for stirring with standard stir bars
- KimCote volumetric flasks are graduated to Class A volumetric tolerances and are designed from ASTM Specification E288
Specifications
| To Contain | |
| Borosilicate Glass (3.3) | |
| Autoclavable | |
| No | |
| Non-sterile | |
| Flat | |
| Plastic Coated |
| 4000 mL | |
| ±1.0 | |
| Reusable | |
| Wide | |
| A | |
| Glass Stopper | |
| 38 |