missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Plastic Safety-Coated Class A Volumetric Flasks with Glass Stopper

Catalog No. p-4411604
Change view
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
100 mL
1000 mL
2000 mL
250 mL
4000 mL
500 mL
Tolerance:
±0.1 mL
±0.2 mL
±0.3 mL
±0.5 mL
±1.0
Catalog No. Capacity (Metric) Tolerance
11417111 100 mL ±0.1 mL
11417112 250 mL ±0.2 mL
11417113 500 mL ±0.2 mL
11417114 1000 mL ±0.3 mL
11417115 2000 mL ±0.5 mL
01183004 4000 mL ±1.0
6 missing translation for 'options'

Catalog No. 11417111

missing translation for 'mfr': DWK Life Sciences KC92812G100

Only null left

Heavy-duty, wide-mouth flasks

KIMAX KimCote protective glassware coating goes beyond traditional coatings. Should a break occur, KimCote will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. It's ultra-clear, extremely durable, autoclavable and resistant to many common laboratory chemicals. KimCote's unique texture also provides a non-slip handling surface, wet or dry. Flasks are graduated to Class A volumetric tolerances and are designed from ASTM Specification E288. Glass stoppers are included.

  • Neck dimensions will easily accommodate a 50 mL pipet
  • Superior strength, durability and heavy uniform walls ensure safety in the lab
  • Flat interior bottoms are ideal for stirring with standard stir bars
  • KimCote volumetric flasks are graduated to Class A volumetric tolerances and are designed from ASTM Specification E288

Specifications

Closure Material Glass
Capacity (Metric) 100 mL
Material Borosilicate Glass
Height (English) 7.086 in.
Height (Metric) 18 cm
Tolerance ±0.1 mL
Graduated Yes
Mouth Style Wide
Closure Size 16