Learn More
DWK Life Sciences Kimble™ KIMAX™ Filling Funnels
Fire-polished edges reduce chipping
$139.89 - $306.43
Specifications
Material Borosilicate Glass
Certifications/Compliance ASTM E1095, Type I, Class A
Length (Metric) Stem 3.5cm
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (Metric)Top Diameter (Metric) Stem Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric)Top Diameter (Metric) Stem Price Quantity & Availability  
103465D
View Documents Promotion Details
DWK Life Sciences
29020125
12.5cm 1.9cm
Pack of 2 Each for $139.89
Case of 6 Pack for $839.34
Only null left
103465A
View Documents Promotion Details
DWK Life Sciences
2902060
6cm 1.3cm
Pack of 6 Each for $305.32
Case of 4 Pack for $1,221.28
Only null left
103465B
View Documents Promotion Details
DWK Life Sciences
2902080
8cm 1.5cm
Pack of 6 Each for $266.25
Case of 4 Pack for $1,065.00
Only null left
103465C
View Documents Promotion Details
DWK Life Sciences
29020100
10cm 1.8cm
Pack of 6 Each for $280.46
Case of 4 Pack for $1,121.84
Only null left
103465E
View Documents Promotion Details
DWK Life Sciences
29020150
15cm 1.9cm
Pack of 2 Each for $306.43
Case of 6 Pack for $1,838.58
Only null left
Description
Description
Fire-polished edges reduce chipping
  • Material: 33 expansion borosilicate glass
  • Fire-polished edges
  • Clear
Specifications
Specifications
Borosilicate Glass
ASTM E1095, Type I, Class A
3.5cm