missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Kimble™ KIMAX™ Distilling-Moisture Receiver

Catalog No. 1031057
Change view
Click to view available options
Standard Taper Joint:
24/40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
1031057 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1031057 Supplier DWK Life Sciences™ Supplier No. K22012 10

With two arm outlets

  • Modified Dean-Stark trap calibrated To Contain with standard taper 24/40 joints and a side arm bridge to equalize pressure
  • 10mL receiver graduated from 0 to 1mL in 0.1mL divisions ±0.05mL, and from 1 to 10mL in 0.2mL divisions ±0.1mL
  • Durable white scale
  • Designed from ASTM™ E 123-92, Style A

Specifications

Includes Two arm outlets
For Use With (Application) Synthetic chemistry to collect water (or other liquid) from a reactor
Type Distilling Moisture Receiver
Capacity 0.33 oz., 10mL
Certifications/Compliance ASTM E123
Capacity (Metric) 10mL
For Use With (Equipment) Reflux condenser
Standard Taper Joint 24/40
Capacity (English) 0.33 oz.