Learn More
DWK Life Sciences Kimble™ KIMAX™ Class A Flasks Unserialized with Snap Cap
Calibrated To Contain
Supplier:  DWK Life Sciences 28008500
 View more versions of this product
Catalog No. 10-041-26P
Only null left
Explore available promotions
Description
Description
  • Meet Class A tolerances
  • Graduation line is blasted on neck
  • Enlarged top of cap will protect neck if flask is tipped over
  • Polyethylene snap cap
Compliance
Designed from ASTM™ Specification E 288, Class A unserialized requirements
Specifications
Specifications
Calibrated To Contain
500 mL
26 cm
Yes
Polyethylene
10.2 in.
±0.2 mL
5
Product Suggestions
Product Suggestions