missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble Chase™ Baume/SG Dual Scale Hydrometers
Available in a variety of graduation ranges and overall lengths
$90.68 - $97.86
Specifications
| Type | Specific Gravity and Baume Hydrometer |
|---|---|
| Includes | Certificate of Traceability |
| Catalog Number | Mfr. No. | Baume Range | Baume Divisions | Specific Gravity Range | Increments | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Baume Range | Baume Divisions | Specific Gravity Range | Increments | Price | Quantity & Availability | ||
|
10310327
|
DWK Life Sciences
521220026 |
0 to 26° | 0.2° | 1.000 to 1.220 | 0.002 |
Each for $96.96
|
|
||
|
10310328
|
DWK Life Sciences
521222443 |
24 to 43° | 0.2° | 1.200 to 1.420 | 0.002 |
Each for $97.86
|
|
||
|
10310331
|
DWK Life Sciences
521220072 |
0 to 72° | 1.0° | 1.000 to 2.000 | 0.01 |
Each for $90.68
|
|
||
Description
- Serial number printed on the scale
- Ballast is made from steel pellets and a binder, and is free of heavier metals
- Includes Certificate of Traceability
- Certificate of Traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards
- Meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy
Specifications
| Specific Gravity and Baume Hydrometer | |
| Certificate of Traceability |