Learn More
DWK Life Sciences Kimble Chase™ Baume/SG Dual Scale Hydrometers
Available in a variety of graduation ranges and overall lengths
Supplier: DWK Life Sciences 521222443
Description
- Serial number printed on the scale
- Ballast is made from steel pellets and a binder, and is free of heavier metals
- Includes Certificate of Traceability
- Certificate of Traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards
- Meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy
Chemical Identifiers
| Specific Gravity and Baume Hydrometer | |
| Certificate of Traceability |
Specifications
| Specific Gravity and Baume Hydrometer | |
| 300 mm | |
| 0.2° | |
| 16 mm | |
| 1.200 to 1.420 |
| 0.002 | |
| Certificate of Traceability | |
| 7 mm | |
| 24 to 43° | |
| 300 mL |