Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials with Unattached PTFE-Faced/White Rubber Lined Closures
Unattached closed top black phenolic closures
Supplier:  DWK Life Sciences 60812B2
 View more versions of this product
Catalog No. 14-957-242
Only null left
Explore available promotions
Description
Description
  • Manufactured from clear 33 expansion borosilicate glass
  • Closures includes PTFE-faced/white rubber liners
Specifications
Specifications
Black
PTFE/Rubber
17 mm
8 mL
2.16 dr.
Screw Cap
ASTM Type I
A
Clear
Borosilicate Glass
Screw
Phenolic
60 mm
15-425
Product Suggestions
Product Suggestions