Learn More
DWK Life Sciences Kimble™ GL 45 Clear Pour Ring
GREENER_CHOICE
Clear Polypropylene Pour Rings for use with GL 45 media bottles and flasks.
Supplier:  DWK Life Sciences 14395C453
 View more versions of this product
Catalog No. 025430
Only null left
Description
Description
Clear Polypropylene Pour Rings for use with GL 45 media bottles and flasks.
  • Solid top polypropylene caps with internal molded seal rings
  • Caps are designed to fit GL 45 threaded Media Bottles and Flasks
  • Autoclavable 140°C
  • Colors available in Gray, Green, Pink, Blue, White and Orange
Specifications
Chemical Identifiers
Polypropylene
140°C
10 Cs.
GL 45
Specifications
Orange
140°C
GL 45
Media bottle
Polypropylene
10 Cs.
Polypropylene Cap
Product Suggestions
Product Suggestions