missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Black Phenolic Screw Caps
Bond between liner and cap retains adhesive properties even after repeated autoclaving
$1281.23 - $1506.37
Specifications
| Color | Black |
|---|---|
| Material | Phenolic Resin |
| For Use With (Equipment) | Bottle |
| Autoclavable | Autoclavable |
| Catalog Number | Mfr. No. | Neck Finish | Certifications/Compliance | No. per Case | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Neck Finish | Certifications/Compliance | No. per Case | Price | Quantity | |||
|
14-930-14G
|
DWK Life Sciences
45066B38 |
38-430 | ASTM E982, Class A | 150 Cs. |
Case of 150 Each for $1,506.37
|
|
|||
|
14-930-15D
|
DWK Life Sciences
45066C13 |
13-415 | ASTM E982, Class B | 300 Cs. |
Case of 300 Each for $1,281.23
|
|
|||
|
14-930-15E
|
DWK Life Sciences
45066C15 |
15-415 | ASTM E982, Class B | 300 Cs. |
Case of 300 Each for $1,281.23
|
|
|||
Description
Bond between liner and cap retains adhesive properties even after repeated autoclaving
With PTFE-faced rubber liner
- Identified in ASTM™ specification E 982-94, Class B requirements
- Has fewer threads than regular GPI caps for easier application and removal
- Not recommended for use with standard GPI finishes usually found on blown-molded bottles
- Identified in ASTM specification E 982-94, Class A requirements
Specifications
| Black | |
| Phenolic Resin | |
| Bottle | |
| Autoclavable |