Learn More
DWK Life Sciences Kimble™ Black Phenolic Screw Caps
Bond between liner and cap retains adhesive properties even after repeated autoclaving
Supplier:  DWK Life Sciences 45066C13
 View more versions of this product
Catalog No. 14-930-15D
Only null left
Explore available promotions
Description
Description
Bond between liner and cap retains adhesive properties even after repeated autoclaving
With PTFE-faced rubber liner
  • Identified in ASTM™ specification E 982-94, Class B requirements
  • With 14B rubber liner (SBR)
    • Has fewer threads than regular GPI caps for easier application and removal
    • Not recommended for use with standard GPI finishes usually found on blown-molded bottles
    • Identified in ASTM specification E 982-94, Class A requirements
    Specifications
    Specifications
    Black
    Phenolic Resin
    13-415
    Yes
    Screw Cap
    0.55 in.
    Phenolic
    Autoclavable
    14mm
    300 Cs.
    ASTM E982, Class B
    14 mm
    Bottle
    PTFE/Rubber
    Phenolic Screw Cap
    Product Suggestions
    Product Suggestions