missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Amber Glass Straight-Sided Jars

Wide-mouth design for efficient addition and removal of contents

$182.32 - $211.21

Specifications

Color Amber
For Use With (Application) Protect Contents from UV Rays and Ideal for Light Sensitive Products
Includes Cap
Material Soda Lime Glass
Packaging Convenience
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
13756795
View Documents Promotion Details
DWK Life Sciences
5420458V21
125 mL
Case of 24 Each for $182.32
Only null left
 
13756797
View Documents Promotion Details
DWK Life Sciences
5420870V21
250 mL
Case of 12 Each for $211.21
Only null left
 
Description

Description

All 32-ounce jars are packed in corrugated cartons with divider cells.

Specifications

Specifications

Amber
Cap
Convenience
Manufactured from Soda Lime Glass Conforming to USP Type III and ASTM E438, Type II Requirements
Round
Pulp/Vinyl
Cap
Protect Contents from UV Rays and Ideal for Light Sensitive Products
Soda Lime Glass
Black
Wide
Straight-sided Jar
Phenolic
Videos
SDS
Documents

Documents