Learn More
DWK Life Sciences Kimble™ Amber Boston Rounds with Taperseal Caps
Narrow-mouth design provides more controlled pouring than wide-mouth.
Supplier:  DWK Life Sciences 5120220C25
 View more versions of this product
Catalog No. 13756577
Only null left
Description
Description
  • Amber colored glass protects contents from UV rays and is ideal for light sensitive products.
Specifications
Chemical Identifiers
Amber
Glass
Black
Screw Cap
Specifications
2 oz., 60 mL
2 oz.
Glass
Black
Taperseal
60 mL
Amber
Bulk Pack/Shrink Module
Boston Round Bottle
Screw Cap
Product Suggestions
Product Suggestions