missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Amber Boston Rounds with 14B White Rubber-lined Caps

Narrow-mouth design provides more controlled pouring than wide-mouth.

Supplier:  DWK Life Sciences 5120220C24

 View more versions of this product

Catalog No. 13756565


Only null left

Description

Description

  • Amber colored glass protects contents from UV rays and is ideal for light sensitive products.

Specifications

Chemical Identifiers

Amber
White
Phenolic
Glass
14B Rubber
Screw Cap

Specifications

2 oz., 60 mL
2 oz.
1.53 in.
White
3.7 in.
Phenolic
39 mm
60 mL
Amber
Glass
94 mm
14B Rubber
20-400 mm
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions