missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Scott™ Essential High Capacity Hard Roll Towels

Catalog No. 19413013
Change view
Click to view available options
For Use With (Equipment):
Kimberly-Clark Professional Dispensers
Universal Hard Roll Towel Dispensers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19413013 Universal Hard Roll Towel Dispensers
296361883 Kimberly-Clark Professional Dispensers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19413013 Supplier Kimberly-Clark Professional™ Supplier No. 01000

Long-lasting roll towel reduces maintenance time, run-outs and complaints. Highly absorbent, use less and waste less. Small cases for easy storage and handling. Compatible with most universal HRT dispensers. Meets EPA standards. FSC™ and EcoLogo™ certified. Elemental chlorine-free bleaching.

  • 12,000 feet per case; 1000 feet per roll; 12 rolls per case
  • Compatible with many Kimberly-Clark Professional hard roll paper towel dispensers, along with many universal hard roll dispensers designed to hold roll paper towels with 1.5 inch cores
  • High capacity: reduce maintenance time, run-outs and complaints with long-lasting bulk paper towels (as compared to a folded towel system)
  • Highly absorbent commercial paper towels let you use less and waste less
  • Bulk paper towels come in small cases for easy storage and handling at your business

Specifications

Product Type Hard Roll Towel
Certifications/Compliance EPA
Color White
For Use With (Equipment) Universal Hard Roll Towel Dispensers