Learn More
Ansell KleenGuard™ A40 Blue Breathable Back Coveralls with Hood
Breathable coveralls with a blue back, from shoulders to ankles, feature a microporous film laminate fabric that provides barrier protection against hazardous particulates, aerosols, and low-hazard liquid splash.
Supplier:  Ansell 37163
 View more versions of this product
Catalog No. 19-048-601
Only null left
Explore available promotions
Description
Description
Breathable coveralls with a blue back, from shoulders to ankles, feature a microporous film laminate fabric that provides barrier protection against hazardous particulates, aerosols, and low-hazard liquid splash.
  • Breathable back, made of SMS fabric, from the shoulders to ankles, which helps keep the wearer cool
  • Each protective suit comes with a zipper front, a hood and elastic wrists and ankles
  • Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates
  • Material gives wearers a better liquid barrier and a better particulate barrier than Dupont Tyvek
  • Pass NFPA 99 for antistatic materials
  • Pass ASTM F1670 testing for penetration of blood and bodily fluids
  • Meet the ANSI/ISEA 101-1996 sizing standards
  • Popular industries using this product include aviation, pharmaceutical manufacturing, wind blade manufacturing, liquid handling, general manufacturing
Specifications
Specifications
General Purpose Coveralls
Unisex
White, Blue
Elastic
Elastic
Elastic
Serged
Non-toxic Particulates, Bloodborne Pathogens
Breathable
Non-woven Microporous Film Laminate Barrier Over SMS
Healthcare, Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Wind Blade Manufacturing, General Manufacturing
X-Large
Microporous Film Laminate
Limited-use
Elastic
Hooded
Front Zipper
Non-sterile
Anti-static
ANSI/ISEA 101-1996, ASTM F1670, ASTM F1671, ASTM F903, NFPA 99
KleenGuard™ A40
Product Suggestions
Product Suggestions