missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A30 Coveralls with Elastic Back, Wrists, Ankles, Attached Hood
Limited-use coveralls designed to protect against hazardous dry particles, aerosols and low-hazardous limited liquid splash.
Supplier: Ansell 46116
Description
- Air permeable SMS fabric incorporates a web of microfibers that filter particulates and water-based liquids
- Suitable for many applications including pharmaceutical research and food processing
- Provides protection from low-hazardous particles and liquids
- Strong and abrasion-resistant
- Suitable for multiple wearings
- Seamless front to provide more protection in the primary exposure areas
- Zipper front with 1 in. (2.5 cm) flap
- Elastic back, wrists, ankles, and attached hood
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Hooded | |
| Front Zipper | |
| Particle, Liquid | |
| NFPA 99 | |
| Pharmaceutical Research, Aerospace, Maintenance, General Manufacturing, Food |
| 3X-Large | |
| SMS Fabric | |
| Limited-use | |
| Elastic | |
| Attached | |
| Non-sterile | |
| Elastic | |
| KleenGuard™ A30 | |
| 21/Cs. |