missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A10 Light-Duty Coveralls
Light-duty coveralls are economically priced for protection in nonhazardous environments.
$116.02 - $126.49
Specifications
| Product Type | Light-duty Coveralls |
|---|---|
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Disposable | Limited-use |
| Catalog Number | Mfr. No. | Description | Size | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Size | Price | Quantity | ||||
|
19-812-878
|
Ansell
10625 |
Without Hood and Boots | 3X-Large |
N/A
|
|
||||
|
19-812-877
|
Ansell
10616 |
Without Hood and Boots | 2X-Large |
N/A
|
|
||||
Description
- Lightweight, breathable fabric keeps workers cool and comfortable
- Zipper front
- Elastic wrists
- Available with or without attached hood and boots
- White
Compliance
Meets ANSI/ISEA 101-1996 standards.
Specifications
| Light-duty Coveralls | |
| Polypropylene | |
| Limited-use | |
| Front Zipper | |
| Non-sterile | |
| KleenGuard™ A10 |
| Unisex | |
| White | |
| Elastic | |
| Serged | |
| ANSI/ISEA 101-1996 | |
| Food Processing, General Maintenance, Manufacturing |