missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ A10 Light-Duty Coveralls
Light-duty coveralls are economically priced for protection in nonhazardous environments.
Supplier: Ansell 10625
Description
- Lightweight, breathable fabric keeps workers cool and comfortable
- Zipper front
- Elastic wrists
- Available with or without attached hood and boots
- White
Compliance
Meets ANSI/ISEA 101-1996 standards.
Specifications
| Light-duty Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Front Zipper | |
| Non-sterile | |
| Without Hood and Boots | |
| Food Processing, General Maintenance, Manufacturing |
| 3X-Large | |
| Polypropylene | |
| Limited-use | |
| Collared | |
| Serged | |
| ANSI/ISEA 101-1996 | |
| KleenGuard™ A10 |