missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech™ Pure G3 Hand-Specific Latex Cleanroom Gloves Multi-Pack

G3 non-sterile latex gloves are 12 in. in length and packaged hand-specific in a multi-pack with 50 left hand and 50 right hand per bag . Recommended for Class 10/ISO 4 or higher cleanroom environments.

$341.38 - $341.38

Specifications

Certifications/Compliance ASTM D3578, IEST-RP-CC005.2
Cleanroom Class Class 10 (ISO 4)
AQL 1.5
Color Natural
Exterior Finish Fully Textured
View More Specs

Alert:

This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
11-390-10A
View Documents
Ansell
56827
6
Case of 200 Pair for $341.38
Only null left
 
11-390-10D
View Documents
Ansell
56830
6.5
Case of 200 Pair for $341.38
Only null left
 
11-390-10C
View Documents
Ansell
56829
7
Case of 200 Pair for $341.38
Only null left
 
11-390-10B
View Documents
Ansell
56828
7.5
Case of 200 Pair for $341.38
Only null left
 
11-390-10E
View Documents
Ansell
56831
8
Case of 200 Pair for $341.38
Only null left
 
11-390-10F
View Documents
Ansell
56832
8.5
Case of 200 Pair for $341.38
Only null left
 
11-390-10G
View Documents
Ansell
56833
9
Case of 200 Pair for $341.38
Only null left
 
Description

Description

  • 100% latex
  • Low residual protein content
  • Textured palms and fingertips for a secure grip
  • Beaded cuff to help prevent roll-down
  • 12 in. length, hand-specific
  • Recommended for ISO Class 4 or higher cleanroom environments
  • Double bagged with case liner
  • Used for medical device manufacturing, pharmaceuticals or research labs
Specifications

Specifications

ASTM D3578, IEST-RP-CC005.2
1.5
Fully Textured
No
Powder Free
7.9 mil
Kimtech™ Pure G3
30.5 cm
Class 10 (ISO 4)
Natural
Specific
Latex
Non-sterile
0.20 mm
12 in.
Videos
SDS
Documents

Documents