missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Jackson Saftey™ G60 Level 2 Cut Resistant Gloves

$76.63 - $76.63

Specifications

Color Black,White
Cut Resistance Rating ANSI Level 2/EN388 Level 3
Material Fiber,Polyurethane
For Use With (Application) Metal fabrication, Glass handling, aerospace and automotive assembly
Series G60
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-005-450 Kimberly-Clark™ Professional
KC38689
Small Case of 12 Each for $76.63
19-005-451 Kimberly-Clark™ Professional
KC38690
Medium Case of 12 Each for $76.63
19-005-453 Kimberly-Clark™ Professional
KC38692
X-Large Case of 12 Each for $76.63
19-005-452 Kimberly-Clark™ Professional
KC38691
Large Case of 12 Each for $76.63
19-005-454 Kimberly-Clark™ Professional
KC38693
2X-Large Case of 12 Each for $76.63
Specifications Description & Specifications

Specifications

Black,White
Fiber,Polyurethane
G60
ANSI Level 2/EN388 Level 3
Metal fabrication, Glass handling, aerospace and automotive assembly