missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ G60 Polyurethane Coated Cut-Resistant Gloves, ANSI Level A2
Description
- Offer EN388 Level 3 / ANSI Level 2 Cut Resistance and CPPT = 686 grams
- Made of ultra-high molecular weight polyethylene (UHMWPE) yarn with nylon and Lycra fiber, which provides both cut resistance and a comfortable fit
- Polyurethane coating enhances grip and the dark color helps hide dirt for longer wear
- Each pair has a dedicated right and left glove
Specifications
Specifications
| Coating Material | Polyurethane |
| Color | Black, White |
| Cut Resistance Rating | ANSI A2 |
| Hand | Specific |
| Latex Free | Yes |
| Material | Dyneema, Fiber, Lycra |
| Product Type | Cut-resistant Gloves |
| Size | 2X-Large |
| EN388 Standard | EN 388 Cut Level 3 |
| For Use With (Application) | Metal fabrication, Glass handling, aerospace and automotive assembly |
| Show More |