Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Available in natural (clear) or choice of four colors
Supplier: DWK Life Sciences 7366518
Description
Available in natural (clear) or choice of four colors
- For plain-end culture tubes
- Keep tube contents sterile without need for cotton plugging
- Polypropylene
- Autoclavable
Chemical Identifiers
| Push Cap | |
| Polypropylene | |
| Reusable |
| Autoclavable | |
| Non-sterile |
Specifications
| Push Cap | |
| Polypropylene | |
| Non-sterile | |
| Reusable |
| Autoclavable | |
| Blue | |
| 18 mm Plain-end Culture Tubes |