missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIFC1 Recombinant Rabbit Monoclonal Antibody (23GB6000)
GREENER_CHOICE

Catalog No. MA552968
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552968 100 μL
MA552969 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552968 Supplier Invitrogen™ Supplier No. MA552968
Only null left

Rabbit Recombinant Monoclonal Antibody

KIFC1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Minus end-directed microtubule-dependent motor required for bipolar spindle formation (PubMed:15843429). May contribute to movement of early endocytic vesicles. Regulates cilium formation and structure. [UniProt].

Specifications

Antigen KIFC1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6000
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Kifc1
Gene Accession No. Q5XI63, Q9BW19
Gene Alias 100042970; EMBL:AAH83827.1, ECO:0000312; Gm4137; HSET; Kifc1; kifc1 {ECO:0000312; Kifc4; Kifc5a; Kifc5b; kinesin family c-terminal 5A; kinesin family member C1; kinesin motor protein KIFC4; kinesin-like 2; kinesin-like 2a; kinesin-like protein 2; Kinesin-like protein KIFC1; kinesin-related protein 1; Kinesin-related protein HSET; kinesin-related protein KIFC5A; kinesin-related protein KRP1; KNSL2; Knsl2a; Krp1; RGD:1359118}; Tctex7; Tctex-7; Tctex7a; Tctex-7A
Gene Symbols Kifc1
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIFC1 (1-69AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294286, 3833
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less