missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIF1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576971 Supplier Invitrogen™ Supplier No. PA576971

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The kinesins constitute a large family of microtubule-dependent motor proteins, which are responsible for the distribution of numerous organelles, vesicles and macromolecular complexes throughout the cell. Individual kinesin members play crucial roles in cell division, intracellular transport and membrane trafficking events including endocytosis and transcytosis. KIF1B is a member of the KIF1/Unc104 family of kinesin-like proteins that are involved in the transport of mitochondria or synaptic vesicles in axons. KIF1B is an amino-terminal-type motor protein that is ubiquitously expressed, with the most abundant levels in differentiated nerve cells. The human KIF1B gene maps to chromosome 1p36. 2. Defects in axonal transport due to mutations at the KIF1B gene can underlie the human peripheral neuropathy phenotype. The mouse KIF1B gene generates an alternatively spliced transcript, which produces two isoforms.

Specifications

Antigen KIF1B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KIF1B
Gene Accession No. O60333, O88658
Gene Alias A530096N05Rik; AI448212; AI506502; CMT2; CMT2A; CMT2A1; D4Mil1e; HMSNII; KIAA0591; KIAA1448; Kif1b; KIF1Bp130; KIF1Bp204; kinesin family member 1B; kinesin heavy chain member 1B; kinesin superfamily protein KIF1B; kinesin-family protein KIF1Bbeta3; kinesin-like protein KIF1B; Klp; N-3 kinesin; NBLST1
Gene Symbols KIF1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KIF1B: 13-120.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117548, 23095
Target Species Human, Rat, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less