missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA0922 Polyclonal Antibody

Description
Immunogen sequence: LNCTLENGVP CVIQESAPVH NSFIDWSATC EGQFSSAYCP LELNDYNAFP EENMNYANGF PCPADVQTDF IDHNSQSTWN TPPNMPAAWG HASFISSP Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 69%.
Specifications
Specifications
| Antigen | KIAA0922 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TMEM131L |
| Gene Accession No. | A2VDJ0 |
| Gene Alias | KIAA0922; TMEM131L; Transmembrane protein 131-like |
| Gene Symbols | TMEM131L |
| Show More |