missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0528 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Orangutan
C2CD5 is required for insulin-stimulated glucose transport and glucose transporter SLC2A4/GLUT4 translocation from intracellular glucose storage vesicle (GSV) to the plasma membrane (PM) in adipocytes. It binds phospholipid membranes in a calcium-dependent manner and is necessary for the optimal membrane fusion between SLC2A4/GLUT4 GSV and the PM.
Specifications
Specifications
| Antigen | KIAA0528 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | C2CD5 |
| Gene Accession No. | Q86YS7 |
| Gene Alias | C2 Calcium-dependent domain containing 5, C2CD5, CDP138, KIAA0528, |
| Gene Symbols | C2CD5 |
| Show More |