missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0460 Goat anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
RPRD2 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleoplasm; RNA polymerase II, holoenzyme; RNA polymerase II complex binding; mRNA 3′-end processing; snRNA transcription by RNA polymerase II.
Specifications
Specifications
| Antigen | KIAA0460 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | RPRD2 |
| Gene Accession No. | Q5VT52 |
| Gene Alias | KIAA0460 |
| Gene Symbols | RPRD2 |
| Show More |