missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0317 Monoclonal Antibody (K56023_20C3), Invitrogen™
Mouse Monoclonal Antibody
$1,557.73 - $2,745.56
Specifications
| Antigen | KIAA0317 |
|---|---|
| Clone | K56023_20C3 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: FDIIVLSEDE KNIVERNVST SGVSIYFEAY LYNATNCSST PWHLPPMHMT SSQRRPSTAV DEEDEDSPSE CHTPEKVKKP
KIAA0317 Monoclonal antibody specifically detects KIAA0317 in Human, Rat samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| KIAA0317 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| apoptosis resistant E3 ubiquitin protein ligase 1; apoptosis-resistant E3 ubiquitin protein ligase 1; Apoptosis-resistant HECT-type E3 ubiquitin transferase 1; AREL1; fibrosis-inducing E3 ligase 1; FIEL1; KIAA0317; protein KIAA0317 | |
| Recombinant human AREL1 (immunogen range is 271-350). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| AREL1 |
| K56023_20C3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| O15033 | |
| 299197, 9870 | |
| IgG2b | |
| Protein A | |
| Antibody |