missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KHK Monoclonal Antibody (K52018_4C12), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555643
Description
Sequence of this protein is as follows: MEEKQILCVG LVVLDVISLV DKYPKEDSEI RCLSQRWQRG GNASNSCTVL SLLGAPCAFM GSMAPGHVAD FLVADFRRRG VDVSQVAWQS KGDTPSSCCI INNSNGNRTI VLHDTSLPDV SATDFEKVDL TQFKWIHIEG RNASEQVKML QRIDAHNTRQ PPEQKIRVSV EVEKPREELF QLFGYGDVVF VSKDVAKHLG FQSAEEALRG LYGRVRKGAV LVCAWAEEGA DALGPDGKLL HSDAFPPPRV VDTLGAGDTF NASVIFSLSQ GRSVQEALRF GCQVAGKKCG LQGFDGIV
KHK encodes the gene ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The splice variant presented encodes the highly active form found in liver. renal cortex. and small intestine. while the alternate variant encodes the lowe.Specifications
| KHK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P50053, Q02974 | |
| KHK | |
| Recombinant human KHK (immunogen range is 1-298). | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K52018_4C12 | |
| Unconjugated | |
| KHK | |
| fructokinase; hepatic fructokinase; KETHPRO; ketohexokinase; ketohexokinase (fructokinase); ketohexokinase isoform a; Khk; testicular tissue protein Li 102 | |
| Mouse | |
| Protein A | |
| RUO | |
| 25659, 3795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |