missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Khellin, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| CAS | 82-02-0 |
| Linear Formula | C14H12O5 |
| MDL Number | MFCD00005007 |
| Melting Point | 150°C to 154°C |
| Boiling Point | 180°C to 200°C (0.05 mmHg) |
| Synonym | 4,9-Dimethoxy-7-methyl-5 H-furo[3,2-g][1 ]benzopyran-5-one |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 260.24 |
| Percent Purity | ≥98.0% (HPLC) |
| Show More |