missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Khellin, 95%

Catalog No. AC461800050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461800050 5 g
AC461800250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461800050 Supplier Thermo Scientific Chemicals Supplier No. 461800050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82-02-0
Molecular Formula C14H12O5
Molecular Weight (g/mol) 260.25
InChI Key HSMPDPBYAYSOBC-UHFFFAOYSA-N
IUPAC Name 4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one
SMILES COC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O

Specifications

Melting Point 150.0°C to 154.0°C
Assay Percent Range 95%
Quantity 5 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 260.24
Chemical Name or Material Khellin
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 201-392-8

RUO – Research Use Only