accessibility menu, dialog, popup

UserName

CAS RN 82-02-0

IUPAC Name: 4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one
Synonyms: kelamin khellin ammi-khellin keloid kellin khellinum bi-kellina quelina khelline I chellina
Molecular Weight (g/mol): 260.25
Molecular Formula: C14H12O5
InChi Key: HSMPDPBYAYSOBC-UHFFFAOYSA-N
SMILES: COC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)