Learn More
Keystone Safety Heavy Duty Polypropylene Cleanroom Lab Coats
Lightweight, latex-free lab coats are made from high quality materials and compatible in Class ISO7 cleanroom environments.
Supplier: Keystone Safety LC0WENWHDXLWHITE
Description
Protect workers against dry particulates, dirt and grime
- Non-woven, nonlinting, spunbond heavyweight polypropylene construction
- Serged seams offer high tensile strength
- Latex-free
- Disposable
- Elastic wrists, snap front, single collar, no pockets
- Available in six sizes
- Flame resistant
Ideal for Class 10,000 environments and above. Commonly used in electronic, medical device and pharmaceutical industries where operator garment control is a concern.
Specifications
| Controlled Environments Lab Coat | |
| Snaps | |
| Single Ply | |
| Electronics, Medical Device, Pharmaceutical |
| White | |
| Single-use | |
| Serged | |
| Class 10,000 (ISO 7) |