missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety Heavy Duty Polypropylene Cleanroom Lab Coats

Catalog No. p-4077905
Catalog No. 19068431 Supplier Keystone Safety Supplier No. LC0WENWHDMEDWHITE

Lightweight, latex-free lab coats are made from high quality materials and compatible in Class ISO7 cleanroom environments.

Protect workers against dry particulates, dirt and grime

  • Non-woven, nonlinting, spunbond heavyweight polypropylene construction
  • Serged seams offer high tensile strength
  • Latex-free
  • Disposable
  • Elastic wrists, snap front, single collar, no pockets
  • Available in six sizes
  • Flame resistant

Ideal for Class 10,000 environments and above. Commonly used in electronic, medical device and pharmaceutical industries where operator garment control is a concern.

Specifications

Product Type Controlled Environments Lab Coat
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Collar Type Single Ply
Seam Type Serged
For Use With (Application) Electronics, Medical Device, Pharmaceutical
Cleanroom Class Class 10,000 (ISO 7)