missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Keyed Alike Safety Lock with Plastic Shackle
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient and cost-effective Safety Lockout Padlocks from Brady.
$66.02 - $269.19
Specifications
| Product Type | Padlock |
|---|---|
| Key Type | Keyed Alike |
| Material | Nylon |
| Diameter (English) Shackle | 0.25 in. |
| Diameter (Metric) Shackle | 6.35 mm |
| Catalog Number | Mfr. No. | Color | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Quantity | Price | Quantity & Availability | ||||
|
19000714
|
Brady™
SDPLRED38PLKA6 |
Red | 6 Pk. |
Pack of 6 Each for $67.37
|
|
||||
|
19000715
|
Brady™
SDPLYLW38PLKA6 |
Yellow | 6 Pk. |
Pack of 6 Each for $67.37
|
|
||||
|
19000716
|
Brady™
SDPLBLU38PLKA6 |
Blue | 6 Pk. |
Pack of 6 Each for $67.37
|
|
||||
|
19000717
|
Brady™
SDPLGRN38PLKA6 |
Green | 6 Pk. |
Pack of 6 Each for $66.02
|
|
||||
|
19000718
|
Brady™
SDPLORG38PLKA6 |
Orange | 6 Pk. |
Pack of 6 Each for $67.37
|
|
||||
|
19000720
|
Brady™
SDPLRED38PLKA12 |
Red | 12 Pk. |
Pack of 12 Each for $135.55
|
|
||||
|
19000722
|
Brady™
SDPLRED38PLKA24 |
Red | 24 Pk. |
Pack of 24 Each for $269.19
|
|
||||
Description
- Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program.
- Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
- Nylon padlocks with plastic shackles are lightweight, non-conductive, non-sparking and ideal for electrical or general lockout applications.
- Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
- Record and retain engraved key codes to streamline replacement of lost keys.
Specifications
| Padlock | |
| Nylon | |
| 6.35 mm | |
| 38.10 mm | |
| 6 |
| Keyed Alike | |
| 0.25 in. | |
| 0.82 x 1.5 x 1.77 in. | |
| Plastic |