missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Keyed Alike Safety Lock with Plastic Shackle
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient and cost-effective Safety Lockout Padlocks from Brady.
Supplier: Brady™ SDPLBLU38PLKA6
Description
- Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program.
- Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
- Nylon padlocks with plastic shackles are lightweight, non-conductive, non-sparking and ideal for electrical or general lockout applications.
- Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
- Record and retain engraved key codes to streamline replacement of lost keys.
Chemical Identifiers
| Padlock | |
| Nylon | |
| 6.35 mm | |
| 38.10 mm | |
| 6 |
| Keyed Alike | |
| 0.25 in. | |
| 0.82 x 1.5 x 1.77 in. | |
| Plastic |
Specifications
| Padlock | |
| Blue | |
| 0.25 in. | |
| 0.82 x 1.5 x 1.77 in. | |
| (6) Padlocks, (6) Keys, (6) English Labels, (6) French Labels, (6) Spanish Labels | |
| 6 |
| Keyed Alike | |
| Nylon | |
| 6.35 mm | |
| 38.10 mm | |
| Plastic | |
| 6 Pk. |