missing translation for 'onlineSavingsMsg'
Learn More

Ketorolac tromethamine, Thermo Scientific™

$138.09 - $404.92

Chemical Identifiers

CAS 74103-07-4
Molecular Formula C19H24N2O6
Molecular Weight (g/mol) 376.41
MDL Number 00887595
InChI Key BWHLPLXXIDYSNW-UHFFFAOYNA-N
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
SMILES NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC469170010
View Documents
Thermo Scientific Chemicals
469170010
1 g
Each for $138.09
Only null left
 
AC469170050
View Documents
Thermo Scientific Chemicals
469170050
5 g
Each for $404.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

74103-07-4
376.41
BWHLPLXXIDYSNW-UHFFFAOYNA-N
NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1
C19H24N2O6
00887595
2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid

Specifications

Glass Bottle
74103-07-4
C19H24N2O6
White to Light Yellow
13-5324
NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1
376.41
Ketorolac tromethamine
Available
Powder
00887595
160°C to 165°C
BWHLPLXXIDYSNW-UHFFFAOYNA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
1 g
Videos
Safety and Handling

Safety and Handling

Skull and crossbones

RTECSNumber : UY7759900

Recommended Storage : Normal conditions

SDS
Documents

Documents