missing translation for 'onlineSavingsMsg'
Learn More

Ketorolac tromethamine, Thermo Scientific™

Catalog No. AC469170050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469170050 5 g
AC469170010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC469170050 Supplier Thermo Scientific Chemicals Supplier No. 469170050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74103-07-4
Molecular Formula C19H24N2O6
Molecular Weight (g/mol) 376.41
MDL Number 00887595
InChI Key BWHLPLXXIDYSNW-UHFFFAOYNA-N
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
SMILES NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Light Yellow
Melting Point 160°C to 165°C
Merck Index 13-5324
Quantity 5 g
Chemical Name or Material Ketorolac tromethamine
Hazard Category Skull and crossbones
RTECSNumber UY7759900
Recommended Storage Normal conditions