missing translation for 'onlineSavingsMsg'
Learn More

Ketorolac Tromethamine, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019314
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019314 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019314 Supplier Merck Emd Millipore Supplier No. PHR1140500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Ketorolac Tromethamine belongs to the class of non-steroidal anti-inflammatory drugs, and its mode of action involves the inhibition of the bodily synthesis of prostagladins. It exhibits anti-inflammatory, antipyretic and analgesic effects.

Specifications

CAS 74103-07-4
Physical Form Neat
Linear Formula C15H13NO3 · C4H11NO3
MDL Number MFCD00887595
Synonym Ketorolac tris salt; (+/-)-5-Benzoyl-2,3-dihydro-1 H-pyrrolizine-1-carboxylic acid tris salt; Toradol
Quantity 500 mg
Formula Weight 376.40
Grade Certified Reference Material