missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole Impurity Standard, MilliporeSigma™ Supelco™

Catalog No. 111001937
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001937 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001937 Supplier Merck Emd Millipore Supplier No. BP1170

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Specifications

CAS 65277-42-1
Linear Formula C26H28 Cl2N4O4
MDL Number MFCD00058579
Synonym Ketoconazole; (+/-)-cis-1-Acetyl-4-(4-[(2-[2,4-dichlorophenyl]-2-[1 H-imidazol-1-ylmethyl]-1,3-dioxolan-4-yl)-methoxy]phenyl)piperazine
Type Pharmacopeia and Metrological Institutes Standards
Quantity 25 mg
Formula Weight 531.43
Grade Pharmaceutical Primary Standard