missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kerosene - Odorless, Ricca Chemical
Supplier: Ricca Chemical Company RSOK01004C
Specifications
| Kerosene (Odorless), Ricca Chemical | |
| 98.0 | |
| Colorless | |
| Amber Glass Bottle | |
| 100% | |
| Liquid |
| 8008-20-6 | |
| 100.0 | |
| 175°C to 325°C | |
| 4 L | |
| Laboratory |
Chemical Identifiers
| 8008-20-6 |
Safety and Handling
Recommended Storage : 15-30°C