missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kerosene - Odorless, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Color | Colorless |
| Boiling Point | 175°C to 325°C |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Percent Purity | 100% |
| Grade | Laboratory |
| Physical Form | Liquid |
| Chemical Name or Material | Kerosene (Odorless), Ricca Chemical |
Safety and Handling
| Recommended Storage | 15-30°C |