missing translation for 'onlineSavingsMsg'
Learn More

Keller's Reagent (Etchant), Ricca Chemical

Catalog No. R4250000100
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4250000100 100 mL
R42500001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R4250000100 Supplier Ricca Chemical Company Supplier No. R4250000100A
Only null left

Specifications

Chemical Name or Material Keller's Reagent
Color Colorless
CAS 7732-18-5, 7697-37-2, 7647-01-0, 7664-39-3
CAS Min % 92.0, 3.62, 1.71, 1.12
CAS Max % 95.0, 3.77, 1.78, 1.16
Synonym Etchant
Grade Laboratory
Packaging Natural Poly Bottle
pH Acidic
Quantity 100 mL
Physical Form Liquid
Show More Show Less