missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KEAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149844
Description
KEAP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.Specifications
| KEAP1 | |
| Polyclonal | |
| Unconjugated | |
| Keap1 | |
| cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 117519, 50868, 9817 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P57790, Q14145, Q9Z2X8 | |
| Keap1 | |
| Recombinant Kelch Like ECH Associated Protein 1 (KEAP1) (Leu327-Cys624). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |