Learn More
Invitrogen™ KCNQ3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA1930
Catalog No. PA1930
Only null left
Description
Description
PA1-930 detects KCNQ3 from mouse, human and rat samples. This antibody is specific for KCNQ3 and does not detect KCNQ1, KCNQ2, KCNQ4 or KCNQ5. PA1-930 has been successfully used in immunofluorescence, immunohistochemistry and immunocytochemistry procedures. Immunohistochemical staining using PA1-930 yielded a strong signal mainly in interneurons and astrocytes in the dentate region of rat hippocampal samples. PA1-930 immunogen is a GST fusion protein encoding the first 71 amino acids of rat KCNQ3.
KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KCNQs (KCNQ1-5) found in the central nervous system and KCNQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.
Specifications
Chemical Identifiers
KCNQ3
-20°C, Avoid Freeze/Thaw Cycles
Polyclonal
Liquid
IgG
Human, Mouse, Rat
KCNQ3
BFNC; BFNC2; EBN2; Kcnq3; KQT-like 3; KV7.3; potassium channel subunit alpha KvLQT3; potassium channel, voltage gated KQT-like subfamily Q, member 3; potassium channel, voltage-gated KQT-like subfamily Q, member 3; potassium channel, voltage-gated, subfamily Q, member 3; potassium voltage-gated channel subfamily KQT member 3; potassium voltage-gated channel subfamily Q member 3; potassium voltage-gated channel, KQT-like subfamily, member 3; potassium voltage-gated channel, subfamily Q, member 3; voltage-gated potassium channel subunit Kv7.3
KCNQ3
Primary
Antigen Affinity Chromatography
1 mg/mL
Immunohistochemistry (Frozen), Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 1mg/mL BSA and 0.02% sodium azide
O43525, O88944, Q8K3F6
110862, 29682, 3786
GST fusion protein encoding the first 71 amino acids of rat KCNQ3.
Antibody
Specifications
KCNQ3
Polyclonal
Unconjugated
KCNQ3
BFNC; BFNC2; EBN2; Kcnq3; KQT-like 3; KV7.3; potassium channel subunit alpha KvLQT3; potassium channel, voltage gated KQT-like subfamily Q, member 3; potassium channel, voltage-gated KQT-like subfamily Q, member 3; potassium channel, voltage-gated, subfamily Q, member 3; potassium voltage-gated channel subfamily KQT member 3; potassium voltage-gated channel subfamily Q member 3; potassium voltage-gated channel, KQT-like subfamily, member 3; potassium voltage-gated channel, subfamily Q, member 3; voltage-gated potassium channel subunit Kv7.3
Rabbit
Antigen Affinity Chromatography
RUO
110862, 29682, 3786
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Frozen), Immunocytochemistry
1 mg/mL
PBS with 1mg/mL BSA and 0.02% sodium azide
O43525, O88944, Q8K3F6
KCNQ3
GST fusion protein encoding the first 71 amino acids of rat KCNQ3.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions