missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNK1 Monoclonal Antibody (5E10A5)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548519
Description
KCNK1 Monoclonal Antibody for Western Blot, Flow, ELISA
This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity. [provided by RefSeq, Jul 2008].Chemical Identifiers
| KCNK1 | |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.05% sodium azide | |
| O00180, Q9Z2T2 | |
| 3775, 59324 | |
| Purified recombinant fragment of human KCNK1 (AA: extra mix) expressed in E. Coli. | |
| Antibody |
| 5E10A5 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Human, Rat | |
| Kcnk1 | |
| AI788889; double-pore potassium channel; DPK; HOHO; HOHO1; Inward rectifying potassium channel protein TWIK-1; K2P1; K2p1.1; Kcnk1; kcnk1 {ECO:0000312; KCNO1; Potassium channel K2P1; potassium channel KCNO1; Potassium channel subfamily K member 1; potassium channel TWIK-1; potassium channel, subfamily K, member 1; potassium channel, two pore domain subfamily K, member 1; potassium inwardly-rectifying channel, subfamily K, member 1; potassium two pore domain channel subfamily K member 1; putative potassium channel TWIK; rabKCNK1; RGD:621447}; rTWIK; tandem of P domains in a weak inward rectifying K+ channel 1; Twik; TWIK1; TWIK-1; two-pore potassium channel; Unknown (protein for MGC:142788) | |
| Kcnk1 | |
| Primary | |
| Protein G |
Specifications
| KCNK1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O00180, Q9Z2T2 | |
| Kcnk1 | |
| Purified recombinant fragment of human KCNK1 (AA: extra mix) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| ELISA, Flow Cytometry, Western Blot | |
| 5E10A5 | |
| Unconjugated | |
| Kcnk1 | |
| AI788889; double-pore potassium channel; DPK; HOHO; HOHO1; Inward rectifying potassium channel protein TWIK-1; K2P1; K2p1.1; Kcnk1; kcnk1 {ECO:0000312; KCNO1; Potassium channel K2P1; potassium channel KCNO1; Potassium channel subfamily K member 1; potassium channel TWIK-1; potassium channel, subfamily K, member 1; potassium channel, two pore domain subfamily K, member 1; potassium inwardly-rectifying channel, subfamily K, member 1; potassium two pore domain channel subfamily K member 1; putative potassium channel TWIK; rabKCNK1; RGD:621447}; rTWIK; tandem of P domains in a weak inward rectifying K+ channel 1; Twik; TWIK1; TWIK-1; two-pore potassium channel; Unknown (protein for MGC:142788) | |
| Mouse | |
| Protein G | |
| RUO | |
| 3775, 59324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |