missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ricca VeriSpec™ KCl Conductivity Standard, Certified Reference Material
ISO 17025/17034 KCl Conductivity Standard Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.
Supplier: Ricca VeriSpec™ RV588750500A
Description
- Potassium Chloride starting materials
- Manufactured and tested in ISO 17025/17034 accredited facilities
- Certified traceable to NIST
- Wide range of concentrations
Specifications
| KCl | |
| Calibration of Conductivity Meter | |
| Approximately 7 | |
| Conductivity Meters and Probes | |
| Conductivity Standard | |
| 500 mL |
| 500μS/cm at 25°C | |
| Colorless | |
| 100°C | |
| Soluble | |
| Certified Reference Material | |
| Certified |