missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCl 0.1D Conductivity Standards, Hach Lange™
Description
- Hach Conductivity Standard 12.85 mS/cm‚ 500 mL‚ S51M002
- KCl 0.1D Conductivity Standard, 500 mL, 0.1 Demal KCl, 12, 85 mS/cm at 25 °C (Radiometer Analytical)
- Each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
- Standard solution verified by Reference Materials Laboratory
- Shelf life of 24 months from production date
Specifications
Specifications
| Methods | Standard solution verified by Reference Materials Laboratory |
| Analyte or Component Names | KCL |
| Conductivity | 12.85mS/cm ± 0.35% at 25°C |
| Physical Form | Liquid |
| Concentration | 12.85 mS/cm ±0.35% at 25°C |
| Element | KCl |
| Quantity | 500 mL |